Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024952.2 | NP_001020123.1 | 10 - 11 | 2054 | 123 | |
GenBank mRNA | AK139648.1 | - | 10 - 11 | 2026 | 123 | |
AK173335.1 | - | 10 - 11 | 2000 | 123 | ||
AY948287.1 | - | 10 - 11 | 1995 | 123 | ||
BC138663.1 | - | 9 - 10 | 1778 | 8861684 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |