Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024952.2 | NP_001020123.1 | 5 - 6 | 1202 | 113 | |
GenBank mRNA | AK139648.1 | - | 5 - 6 | 1174 | 113 | |
AK173335.1 | - | 5 - 6 | 1148 | 113 | ||
AY948287.1 | - | 5 - 6 | 1143 | 113 | ||
BC138663.1 | - | 4 - 5 | 926 | 8861684 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |