Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100026233.mm.1 |
NM_021400.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026233.mm.1 |
NM_021400.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026233.mm.1 |
TC0100003240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026234.mm.1 |
NM_021400.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026234.mm.1 |
NM_021400.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026234.mm.1 |
TC0100003240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026232.mm.1 |
NM_021400.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026232.mm.1 |
NM_021400.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026232.mm.1 |
TC0100003240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026235.mm.1 |
NM_001110146.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026235.mm.1 |
NM_001110146.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026235.mm.1 |
NM_021400.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026235.mm.1 |
NM_021400.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026235.mm.1 |
TC0100003240.mm.1 |
MTA TC |