Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100011201.mm.1 |
NM_026876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011201.mm.1 |
NM_026876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011201.mm.1 |
TC0100001353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011206.mm.1 |
NM_026876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011206.mm.1 |
NM_026876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011206.mm.1 |
TC0100001353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011202.mm.1 |
NM_026876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011202.mm.1 |
NM_026876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011202.mm.1 |
TC0100001353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011203.mm.1 |
NM_026876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011203.mm.1 |
NM_026876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011203.mm.1 |
TC0100001353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011204.mm.1 |
NM_026876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011204.mm.1 |
NM_026876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011204.mm.1 |
TC0100001353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011200.mm.1 |
NM_026876.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011200.mm.1 |
NM_026876.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011200.mm.1 |
TC0100001353.mm.1 |
MTA TC |