Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Allc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053156.2 | NP_444386.2 | 9 - 10 | 928 | 67 | |
GenBank mRNA | AF278712.1 | - | 9 - 10 | 936 | 67 | |
AK005893.1 | - | 9 - 10 | 855 | 67 | ||
BC108957.1 | - | 9 - 10 | 928 | 40046116 | 67 | |
BC108958.1 | - | 9 - 10 | 928 | 40046117 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |