Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161406.1 | NP_001154878.1 | 8 - 9 | 1256 | 64 | |
NM_145890.2 | NP_665897.2 | 6 - 7 | 888 | 64 | ||
GenBank mRNA | AF411211.1 | - | 6 - 7 | 888 | 64 | |
AF411212.1 | - | 8 - 9 | 1227 | 64 | ||
AK076307.1 | - | 8 - 9 | 1242 | 64 | ||
AK161048.1 | - | 8 - 9 | 1138 | 64 | ||
BC013080.1 | - | 8 - 9 | 1196 | 3499860 | 64 | |
BC037233.1 | - | 8 - 9 | 1248 | 4951512 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |