Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289462.1 | NP_001276391.1 | 19 - 20 | 2361 | 147 | |
NM_001289463.1 | NP_001276392.1 | 19 - 20 | 2145 | 147 | ||
NM_008604.4 | NP_032630.2 | 19 - 20 | 1995 | 147 | ||
GenBank mRNA | AK031446.1 | - | 19 - 20 | 1996 | 147 | |
AK033824.1 | - | 19 - 20 | 2056 | 147 | ||
AK076908.1 | - | 19 - 20 | 2209 | 147 | ||
BC034092.1 | - | 19 - 20 | 2017 | 4483558 | 147 | |
BC066840.1 | - | 18 - 19 | 1904 | 147 | ||
M81591.1 | - | 18 - 19 | 1907 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |