Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289462.1 | NP_001276391.1 | 7 - 8 | 1103 | 136 | |
NM_001289463.1 | NP_001276392.1 | 7 - 8 | 887 | 136 | ||
NM_008604.4 | NP_032630.2 | 7 - 8 | 737 | 136 | ||
GenBank mRNA | AK031446.1 | - | 7 - 8 | 738 | 136 | |
AK033824.1 | - | 7 - 8 | 798 | 136 | ||
AK040730.1 | - | 7 - 8 | 774 | 136 | ||
AK052088.1 | - | 6 - 7 | 624 | 136 | ||
AK076908.1 | - | 7 - 8 | 951 | 136 | ||
AK080202.1 | - | 7 - 8 | 765 | 136 | ||
BC034092.1 | - | 7 - 8 | 759 | 4483558 | 136 | |
BC066840.1 | - | 6 - 7 | 646 | 136 | ||
M81591.1 | - | 6 - 7 | 649 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |