Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081078.1 | NP_001074547.1 | 10 - 11 | 4497 | 100 | |
GenBank mRNA | AK158042.1 | - | 4 - 5 | 2599 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |