Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122675.1 | NP_001116147.1 | 4 - 5 | 1568 | 108 | |
NM_001122676.1 | NP_001116148.1 | 4 - 5 | 1568 | 108 | ||
GenBank mRNA | AK046562.1 | - | 4 - 5 | 1569 | 108 | |
AK173248.1 | - | 4 - 5 | 1285 | 108 | ||
BC035528.1 | - | 1 - 2 | 58 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |