Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081307.1 | NP_001074776.1 | 21 - 22 | 2895 | 111 | |
GenBank mRNA | AK039445.1 | - | 9 - 10 | 956 | 111 | |
BC145746.1 | - | 21 - 22 | 2806 | 40131049 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |