Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mroh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281466.1 | NP_001268395.1 | 16 - 17 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |