Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mroh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281466.1 | NP_001268395.1 | 34 - 35 | 4212 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |