Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007842.2 | NP_031868.2 | 12 - 13 | 1484 | 61 | |
GenBank mRNA | AK135603.1 | - | 7 - 8 | 799 | 61 | |
AK141824.1 | - | 12 - 13 | 1479 | 61 | ||
AK148423.1 | - | 12 - 13 | 1480 | 61 | ||
AY512925.1 | - | 8 - 9 | 940 | 61 | ||
U91922.1 | - | 12 - 13 | 1441 | 61 | ||
U92080.1 | - | 1 - 2 | 179 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |