Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243223.1 | NP_001230152.1 | 13 - 14 | 2385 | 100 | |
GenBank mRNA | AK029630.1 | - | 4 - 5 | 492 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |