Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173392.4 | NP_775568.1 | 2 - 3 | 216 | 84 | |
GenBank mRNA | AK045090.1 | - | 2 - 3 | 274 | 84 | |
AK122241.1 | - | 2 - 3 | 197 | 84 | ||
AK170435.1 | - | 2 - 3 | 235 | 84 | ||
BC042669.1 | - | 2 - 3 | 214 | 4235024 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |