Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081082.2 | NP_001074551.1 | 9 - 10 | 1263 | 107 | |
GenBank mRNA | AK008000.1 | - | 3 - 4 | 353 | 107 | |
BC119800.1 | - | 9 - 10 | 1191 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |