Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177129.5 | NP_796103.2 | 14 - 15 | 2126 | 85 | |
GenBank mRNA | AK147573.1 | - | 13 - 14 | 2254 | 85 | |
AK147602.1 | - | 14 - 15 | 2127 | 85 | ||
AK147658.1 | - | 13 - 14 | 2254 | 85 | ||
AK147673.1 | - | 13 - 14 | 2279 | 85 | ||
BC053033.1 | - | 3 - 4 | 334 | 85 | ||
BC066106.1 | - | 14 - 15 | 2046 | 30362931 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |