Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022881.4 | NP_075019.1 | 3 - 4 | 468 | 118 | |
GenBank mRNA | AB042807.1 | - | 3 - 4 | 483 | 118 | |
AF302685.1 | - | 3 - 4 | 474 | 118 | ||
AK036407.1 | - | 3 - 4 | 469 | 118 | ||
BC099387.1 | - | 3 - 4 | 463 | 30846527 | 118 | |
BC132544.1 | - | 3 - 4 | 469 | 40130821 | 118 | |
BC132546.1 | - | 3 - 4 | 469 | 40130823 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |