Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161406.1 | NP_001154878.1 | 1 - 2 | 175 | 68 | |
GenBank mRNA | AF411212.1 | - | 1 - 2 | 146 | 68 | |
AK076307.1 | - | 1 - 2 | 160 | 68 | ||
AK161048.1 | - | 1 - 2 | 57 | 68 | ||
AK167035.1 | - | 1 - 2 | 175 | 68 | ||
BC013080.1 | - | 1 - 2 | 115 | 3499860 | 68 | |
BC037233.1 | - | 1 - 2 | 167 | 4951512 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |