Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205391.1 | NP_001192320.1 | 15 - 16 | 1667 | 114 | |
NM_001205392.1 | NP_001192321.1 | 13 - 14 | 1562 | 114 | ||
NM_029846.4 | NP_084122.2 | 14 - 15 | 1619 | 114 | ||
GenBank mRNA | AB087879.1 | - | 13 - 14 | 1373 | 114 | |
AB087880.1 | - | 14 - 15 | 1430 | 114 | ||
AB087881.1 | - | 15 - 16 | 1478 | 114 | ||
AB476646.1 | - | 15 - 16 | 1478 | 114 | ||
AK005181.1 | - | 14 - 15 | 1836 | 114 | ||
AK030983.1 | - | 14 - 15 | 1560 | 114 | ||
AK131120.1 | - | 12 - 13 | 2649 | 114 | ||
AK155358.1 | - | 13 - 14 | 1498 | 114 | ||
AK170073.1 | - | 13 - 14 | 1496 | 114 | ||
BC049122.1 | - | 15 - 16 | 1592 | 6813377 | 114 | |
BC052087.1 | - | 2 - 3 | 517 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |