Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110212.2 | NP_001103682.1 | 1 - 2 | 94 | 73 | |
NM_146112.4 | NP_666224.3 | 1 - 2 | 94 | 73 | ||
GenBank mRNA | AK034426.1 | - | 1 - 2 | 63 | 73 | |
AK079246.1 | - | 1 - 2 | 64 | 73 | ||
AK144058.1 | - | 1 - 2 | 56 | 73 | ||
AK160878.1 | - | 1 - 2 | 59 | 73 | ||
BC076610.1 | - | 1 - 2 | 40 | 73 | ||
BC089036.1 | - | 1 - 2 | 50 | 30695300 | 73 | |
BY117987.1 | - | 1 - 2 | 94 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |