Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110212.2 | NP_001103682.1 | 8 - 9 | 738 | 73 | |
NM_146112.4 | NP_666224.3 | 8 - 9 | 738 | 73 | ||
GenBank mRNA | AK079246.1 | - | 8 - 9 | 708 | 73 | |
AK144058.1 | - | 8 - 9 | 700 | 73 | ||
AY176043.1 | - | 8 - 9 | 684 | 73 | ||
BC021370.1 | - | 8 - 9 | 705 | 73 | ||
BC076610.1 | - | 8 - 9 | 684 | 73 | ||
BC089036.1 | - | 8 - 9 | 694 | 30695300 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |