Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028916.4 | NP_083192.2 | 7 - 8 | 1199 | 93 | |
GenBank mRNA | AK029501.1 | - | 7 - 8 | 1165 | 93 | |
BC120659.1 | - | 7 - 8 | 1134 | 40129582 | 93 | |
BC125625.1 | - | 7 - 8 | 1134 | 40130140 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |