Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009481.2 | NP_033507.2 | 14 - 15 | 2294 | 85 | |
GenBank mRNA | AK028443.1 | - | 14 - 15 | 2294 | 85 | |
AK049283.1 | - | 15 - 16 | 2407 | 85 | ||
DQ086491.1 | - | 14 - 15 | 2255 | 85 | ||
U67874.1 | - | 14 - 15 | 2255 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |