Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009481.2 | NP_033507.2 | 21 - 22 | 3543 | 109 | |
GenBank mRNA | AK053279.1 | - | 1021 | 109 | ||
DQ086491.1 | - | 21 - 22 | 3504 | 109 | ||
U67874.1 | - | 21 - 22 | 3504 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mus musculus Probe spans exons |