Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018864.5 | NP_061352.2 | 1 - 2 | 87 | ||
GenBank mRNA | AF042730.1 | - | 1 - 2 | 60 | 87 | |
AK078406.1 | - | 1 - 2 | 87 | |||
AK083238.1 | - | 1 - 2 | 87 | |||
AK145472.1 | - | 87 | ||||
AK147778.1 | - | 1 - 2 | 87 | |||
AK152774.1 | - | 1 - 2 | 87 | |||
AK153457.1 | - | 1 - 2 | 87 | |||
AK160203.1 | - | 1 - 2 | 87 | |||
AK160467.1 | - | 1 - 2 | 87 | |||
AK165977.1 | - | 1 - 2 | 87 | |||
AK168457.1 | - | 1 - 2 | 87 | |||
BC049080.1 | - | 1 - 2 | 6419835 | 87 | ||
BY283562.1 | - | 1 - 2 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |