Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173753.4 | NP_776114.2 | 3 - 4 | 471 | 120 | |
GenBank mRNA | AK087849.1 | - | 3 - 4 | 1125 | 120 | |
AK122574.1 | - | 3 - 4 | 419 | 120 | ||
BC079892.1 | - | 3 - 4 | 461 | 30543296 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |