Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025346.1 | NP_079622.1 | 3 - 4 | 640 | 56 | |
GenBank mRNA | AK075599.1 | - | 3 - 4 | 640 | 56 | |
BC016075.1 | - | 3 - 4 | 494 | 4485143 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |