Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048061.1 | NP_001041526.1 | 6 - 7 | 1018 | 69 | |
GenBank mRNA | AK013709.1 | - | 984 | 69 | ||
AK165609.1 | - | 7 - 8 | 965 | 69 | ||
AK182045.1 | - | 2 - 3 | 122 | 69 | ||
AY137376.1 | - | 2 - 3 | 135 | 69 | ||
BC043069.1 | - | 6 - 7 | 972 | 6402930 | 69 | |
BQ445737.1 | - | 3 - 4 | 378 | 69 | ||
DQ397821.1 | - | 5 - 6 | 780 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |