Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF033566.1 | - | 7 - 8 | 893 | 83 | |
AK013974.2 | - | 5 - 6 | 648 | 83 | ||
AY653308.1 | - | 6 - 7 | 783 | 83 | ||
BC002220.1 | - | 3 - 4 | 449 | 3490616 | 83 | |
BC012675.1 | - | 7 - 8 | 915 | 3982920 | 83 | |
CT010293.1 | - | 6 - 7 | 815 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |