Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045520.3 | NP_001038985.2 | 5 - 6 | 710 | 73 | |
GenBank mRNA | AB093212.1 | - | 4 - 5 | 1797 | 73 | |
AK143544.1 | - | 5 - 6 | 711 | 73 | ||
AK168991.1 | - | 5 - 6 | 676 | 73 | ||
BC004080.1 | - | 2 - 3 | 248 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |