Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF128214.1 | - | 2 - 3 | 121 | 105 | |
AF128215.1 | - | 2 - 3 | 121 | 105 | ||
AK162981.1 | - | 2 - 3 | 145 | 105 | ||
BC103608.1 | - | 2 - 3 | 127 | 40046200 | 105 | |
BC103609.1 | - | 2 - 3 | 127 | 40046202 | 105 | |
BC103610.1 | - | 2 - 3 | 127 | 40046204 | 105 | |
BC103614.1 | - | 2 - 3 | 127 | 40046206 | 105 | |
BY154173.1 | - | 2 - 3 | 145 | 105 | ||
M86671.1 | - | 2 - 3 | 122 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |