Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148673.3 | NP_683514.2 | 11 - 12 | 1173 | 100 | |
NM_198936.1 | NP_945174.1 | 10 - 11 | 1094 | 100 | ||
GenBank mRNA | AK029117.1 | - | 10 - 11 | 1064 | 100 | |
AK046262.1 | - | 10 - 11 | 1065 | 100 | ||
AK047597.1 | - | 11 - 12 | 1161 | 100 | ||
AK049178.1 | - | 10 - 11 | 1063 | 100 | ||
BC013810.1 | - | 8 - 9 | 693 | 100 | ||
BC060954.1 | - | 10 - 11 | 1062 | 100 | ||
BC082780.1 | - | 11 - 12 | 1153 | 100 | ||
BC106099.1 | - | 10 - 11 | 1075 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |