Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007799.3 | NP_031825.2 | 8 - 9 | 1298 | 73 | |
GenBank mRNA | AK143581.1 | - | 7 - 8 | 1000 | 73 | |
AK145875.1 | - | 8 - 9 | 1155 | 73 | ||
AK157907.1 | - | 7 - 8 | 1020 | 73 | ||
AK165271.1 | - | 7 - 8 | 966 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Mus musculus Probe spans exons |