Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008795.2 | NP_032821.1 | 11 - 12 | 1301 | 69 | |
GenBank mRNA | AK004998.1 | - | 11 - 12 | 1245 | 69 | |
AK171569.1 | - | 11 - 12 | 1082 | 69 | ||
BC119617.1 | - | 12 - 13 | 1177 | 40061728 | 69 | |
BC119618.1 | - | 6 - 7 | 568 | 69 | ||
X69026.1 | - | 11 - 12 | 1318 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |