Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007963.2 | NP_031989.2 | 9 - 10 | 2057 | 86 | |
GenBank mRNA | AJ001482.1 | - | 8 - 9 | 2212 | 86 | |
AK033517.1 | - | 9 - 10 | 1491 | 86 | ||
BC076620.1 | - | 9 - 10 | 2009 | 86 | ||
BC139762.1 | - | 7 - 8 | 1175 | 40109903 | 86 | |
JQ665270.1 | - | 7 - 8 | 2177 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |