Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175386.3 | NP_780595.1 | 1 - 2 | 227 | 110 | |
GenBank mRNA | AK159126.1 | - | 1 - 2 | 216 | 110 | |
AK159549.1 | - | 1 - 2 | 226 | 110 | ||
BC052079.1 | - | 1 - 2 | 227 | 6402782 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |