Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300000838.mm.1 |
NM_001164056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000838.mm.1 |
NM_001164056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000838.mm.1 |
NM_008875.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000838.mm.1 |
NM_008875.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000838.mm.1 |
TC0300000168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000837.mm.1 |
NM_001164056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000837.mm.1 |
NM_001164056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000837.mm.1 |
NM_008875.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000837.mm.1 |
NM_008875.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000837.mm.1 |
TC0300000168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000839.mm.1 |
NM_001164056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000839.mm.1 |
NM_001164056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000839.mm.1 |
NM_008875.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000839.mm.1 |
NM_008875.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000839.mm.1 |
TC0300000168.mm.1 |
MTA TC |