Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008839.2 | NP_032865.2 | 12 - 13 | 2075 | 129 | |
GenBank mRNA | AK051885.1 | - | 12 - 13 | 1708 | 129 | |
BC089038.1 | - | 12 - 13 | 2075 | 30692343 | 129 | |
BC130228.1 | - | 12 - 13 | 2024 | 40141870 | 129 | |
U03279.1 | - | 12 - 13 | 2020 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |