Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024200.4 | NP_077162.2 | 6 - 7 | 872 | 65 | |
GenBank mRNA | AK046313.1 | - | 6 - 7 | 825 | 65 | |
AK141611.1 | - | 6 - 7 | 873 | 65 | ||
AK220169.1 | - | 6 - 7 | 860 | 65 | ||
AY174062.1 | - | 5 - 6 | 1001 | 65 | ||
BC008137.1 | - | 2 - 3 | 139 | 65 | ||
BC056641.1 | - | 6 - 7 | 831 | 5097345 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |