Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011020.3 | NP_035150.3 | 9 - 10 | 1207 | 111 | |
GenBank mRNA | AB001926.1 | - | 9 - 10 | 1173 | 111 | |
AK144225.1 | - | 9 - 10 | 1244 | 111 | ||
AK163459.1 | - | 10 - 11 | 1379 | 111 | ||
BC012712.1 | - | 9 - 10 | 1276 | 4190161 | 111 | |
BC057002.1 | - | 9 - 10 | 1207 | 5702602 | 111 | |
BC110662.1 | - | 9 - 10 | 1227 | 4511154 | 111 | |
D49482.1 | - | 9 - 10 | 1201 | 111 | ||
U23921.1 | - | 9 - 10 | 1324 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |