Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028140.4 | NP_082416.2 | 4 - 5 | 592 | 70 | |
GenBank mRNA | AK013144.1 | - | 4 - 5 | 484 | 70 | |
AK032080.1 | - | 4 - 5 | 613 | 70 | ||
AK035291.1 | - | 4 - 5 | 555 | 70 | ||
AK077957.1 | - | 4 - 5 | 593 | 70 | ||
BC010483.1 | - | 4 - 5 | 441 | 70 | ||
BC113183.1 | - | 4 - 5 | 523 | 40061891 | 70 | |
BC113790.1 | - | 4 - 5 | 523 | 40061889 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |