Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167748.1 | NP_001161220.1 | 3 - 4 | 605 | 87 | |
NM_029412.1 | NP_083688.1 | 3 - 4 | 605 | 87 | ||
GenBank mRNA | AK018073.1 | - | 3 - 4 | 608 | 87 | |
AK032017.1 | - | 3 - 4 | 449 | 87 | ||
BC117810.1 | - | 3 - 4 | 411 | 40092604 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |