Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286865.1 | NP_001273794.1 | 4 - 5 | 586 | 67 | |
NM_011275.3 | NP_035405.2 | 4 - 5 | 586 | 67 | ||
GenBank mRNA | AF048993.1 | - | 4 - 5 | 547 | 67 | |
AK088032.1 | - | 2 - 3 | 1101 | 67 | ||
BC019411.1 | - | 4 - 5 | 515 | 5133942 | 67 | |
BM943446.1 | - | 1 - 2 | 106 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |