Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039511.1 | NP_001034600.1 | 6 - 7 | 1038 | 137 | |
NM_054102.2 | NP_473443.2 | 6 - 7 | 1038 | 137 | ||
GenBank mRNA | AB055737.1 | - | 6 - 7 | 1034 | 137 | |
AB055738.1 | - | 6 - 7 | 1034 | 137 | ||
AK007291.1 | - | 6 - 7 | 988 | 137 | ||
AK129229.1 | - | 6 - 7 | 976 | 137 | ||
AK144619.1 | - | 6 - 7 | 1017 | 137 | ||
AK145071.1 | - | 6 - 7 | 1029 | 137 | ||
AK145845.1 | - | 6 - 7 | 988 | 137 | ||
AK146001.1 | - | 6 - 7 | 1019 | 137 | ||
AK146494.1 | - | 6 - 7 | 991 | 137 | ||
AK146645.1 | - | 6 - 7 | 989 | 137 | ||
AK159254.1 | - | 6 - 7 | 1020 | 137 | ||
BC004092.1 | - | 5 - 6 | 738 | 3593827 | 137 | |
BC040250.1 | - | 6 - 7 | 1030 | 137 | ||
DQ914523.1 | - | 6 - 7 | 960 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |