Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013730.4 | NP_038758.2 | 3 - 4 | 807 | 73 | |
GenBank mRNA | AB196796.1 | - | 3 - 4 | 712 | 73 | |
AB196797.1 | - | 3 - 4 | 712 | 73 | ||
AB196798.1 | - | 3 - 4 | 712 | 73 | ||
AB196799.1 | - | 3 - 4 | 712 | 73 | ||
AF149791.1 | - | 3 - 4 | 772 | 73 | ||
AF149792.1 | - | 3 - 4 | 772 | 73 | ||
AF160990.1 | - | 3 - 4 | 778 | 73 | ||
AK016818.1 | - | 2 - 3 | 481 | 73 | ||
AK020012.1 | - | 3 - 4 | 808 | 73 | ||
AK036429.1 | - | 3 - 4 | 783 | 73 | ||
AK037780.1 | - | 3 - 4 | 785 | 73 | ||
AK156445.1 | - | 3 - 4 | 809 | 73 | ||
AK169547.1 | - | 1 - 2 | 88 | 73 | ||
AK172138.1 | - | 3 - 4 | 785 | 73 | ||
BC117095.1 | - | 3 - 4 | 748 | 40126414 | 73 | |
BC117099.1 | - | 3 - 4 | 748 | 40126418 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |