Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130174.2 | NP_001123646.1 | 13 - 14 | 865 | 73 | |
NM_001130175.2 | NP_001123647.4 | 13 - 14 | 861 | 73 | ||
NM_001130176.1 | NP_001123648.1 | 12 - 13 | 761 | 73 | ||
NM_001130177.2 | NP_001123649.1 | 12 - 13 | 853 | 73 | ||
NM_001130178.2 | NP_001123650.1 | 13 - 14 | 844 | 73 | ||
NM_001130179.2 | NP_001123651.4 | 13 - 14 | 840 | 73 | ||
NM_001130180.2 | NP_001123652.1 | 12 - 13 | 832 | 73 | ||
NM_001130181.2 | NP_001123653.4 | 12 - 13 | 828 | 73 | ||
NM_011619.3 | NP_035749.1 | 11 - 12 | 823 | 73 | ||
GenBank mRNA | AK146945.1 | - | 13 - 14 | 810 | 73 | |
AK168393.1 | - | 12 - 13 | 798 | 73 | ||
AK169061.1 | - | 13 - 14 | 790 | 73 | ||
BC063753.1 | - | 13 - 14 | 841 | 30293959 | 73 | |
L47549.1 | - | 13 - 14 | 758 | 73 | ||
L47550.1 | - | 13 - 14 | 743 | 73 | ||
L47551.1 | - | 12 - 13 | 744 | 73 | ||
L47552.1 | - | 12 - 13 | 753 | 73 | ||
L47553.1 | - | 12 - 13 | 760 | 73 | ||
L47599.1 | - | 13 - 14 | 764 | 73 | ||
L47600.1 | - | 11 - 12 | 752 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |