Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023200.2 | NP_075689.1 | 6 - 7 | 617 | 157 | |
GenBank mRNA | AF222867.1 | - | 6 - 7 | 617 | 157 | |
AK009342.1 | - | 6 - 7 | 632 | 157 | ||
AK009870.1 | - | 6 - 7 | 610 | 157 | ||
AK031983.1 | - | 1410 | 158 | |||
AK129024.1 | - | 6 - 7 | 634 | 157 | ||
AK145136.1 | - | 6 - 7 | 623 | 157 | ||
AK162517.1 | - | 6 - 7 | 625 | 157 | ||
BC013524.1 | - | 6 - 7 | 618 | 4237290 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |