Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023200.2 | NP_075689.1 | 8 - 9 | 831 | 155 | |
GenBank mRNA | AF222867.1 | - | 8 - 9 | 831 | 155 | |
AK009342.1 | - | 8 - 9 | 846 | 155 | ||
AK031983.1 | - | 1624 | 155 | |||
AK129024.1 | - | 8 - 9 | 848 | 155 | ||
AK145136.1 | - | 8 - 9 | 837 | 155 | ||
AK162517.1 | - | 8 - 9 | 839 | 155 | ||
BC013524.1 | - | 8 - 9 | 832 | 4237290 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |